Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on carbon paste, Type 755

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 10% on carbon paste, Type 755
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3727706

  SDS
alfa aesar™
H3727706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3727714

  SDS
alfa aesar™
H3727714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3727722

  SDS
alfa aesar™
H3727722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solid
Chemical Name or Material Palladium
MDL Number MFCD03457879
Quantity 5g
TSCA TSCA
Name Note 10% on carbon paste, Type 755
Odor Odorless
Color Dark gray
CAS 7440-05-3