missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on activated carbon powder, Type 87L, standard, reduced, nominally 50% water wet

Manufacturer:  Alfa Aesar™ 4592406

 View more versions of this product

Catalog No. AA4592406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Palladium
7440-05-3
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Powder
Black
10% on activated carbon powder, Type 87L, Standard, Reduced, Nominally 50% water wet
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
900 to 1100m2/g
5g

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents