missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on activated carbon powder, Type 58, standard, reduced, nominally 50% water wet

Manufacturer:  Alfa Aesar™ 4435006

 View more versions of this product

Catalog No. AA4435006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic nitrites to 1° amines
Specifications

Specifications

Palladium
7440-05-3
MFCD03457879
Insoluble in water.
[Pd]
106.42
CHEBI:33363
Odorless
5g
10% on activated carbon powder, Type 58, Standard, Reduced, Nominally 50% water wet
Pd
black
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Powder
Black

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents