Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on activated carbon paste, Type 487

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 10% on activated carbon paste, Type 487
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3695806

  SDS
alfa aesar™
H3695806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3695814

  SDS
alfa aesar™
H3695814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3695822

  SDS
alfa aesar™
H3695822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Odor Odorless
Name Note 10% on activated carbon paste, Type 487
Physical Form Solid
MDL Number MFCD03457879
CAS 7440-05-3
Color Dark gray
Quantity 5g