missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on activated carbon paste, Type 487

Manufacturer:  Alfa Aesar™ H3695814

 View more versions of this product

Catalog No. AAH3695814



Specifications

Specifications

Palladium
7440-05-3
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Solid
Gray
10% on activated carbon paste, Type 487
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
Odorless
25g

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents