Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on activated carbon paste, A402028-10

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 10% on activated carbon paste, A402028-10
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3830306

  SDS
alfa aesar™
3830306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830314

  SDS
alfa aesar™
3830314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830318

  SDS
alfa aesar™
3830318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 10% on activated carbon paste, A402028-10
MDL Number MFCD03457879
CAS 7440-05-3
Color Black
Quantity 5g