missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 1% on granular carbon, reduced

Manufacturer:  Alfa Aesar™ 4646222

 View more versions of this product

Catalog No. AA4646222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Palladium
7440-05-3
MFCD00011167
[Pd]
106.42
CHEBI:33363
100g
1% on granular carbon, Reduced
Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Pellet
3mm (0.13 in.)

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
CHEBI:33363
[Pd]
Pd
MFCD00011167
23938
palladium
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents