Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 1% on activated carbon powder, standard, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 1% on activated carbon powder, Standard, reduced, nominally 50% water wet
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3829209

  SDS
alfa aesar™
3829209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3829218

  SDS
alfa aesar™
3829218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
Name Note 1% on activated carbon powder, Standard, reduced, nominally 50% water wet
TSCA TSCA
MDL Number MFCD03457879
Quantity 10g
Weight 900 to 1100m2/g