Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 1% on activated carbon powder, eggshell, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 1% on activated carbon powder, nominally 50% water wet, Eggshell, Reduced
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3852706

  SDS
alfa aesar™
3852706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3852722

  SDS
alfa aesar™
3852722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3852714

  SDS
alfa aesar™
3852714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD03457879
Weight 900 to 1100m2/g
Chemical Name or Material Palladium
TSCA TSCA
Quantity 5g
Name Note 1% on activated carbon powder, nominally 50% water wet, Eggshell, Reduced