Please login to your online account to display your discounted pricing

Palladium on 1/8-inch alumina pellets, 0.5% Pd, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC195080010

 View more versions of this product

Catalog No. AC195080010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Additional Information Loss on abrasion: 3.5%
Chemical Name or Material Palladium on 1/8-inch alumina pellets
Quantity 1kg
CAS 7440-05-3
Health Hazard 1
Density 700g/L (tapped)
Physical Form Pellets
Assay 0.5% (Pd)
Molecular Formula Pd
Loss on Drying 1% (typically)
Formula Weight 106.42
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Packaging Glass bottle
Pore Volume 0.54mL/g
Specific Surface Area 230m2/g
Color Gray

Specifications

Chemical Name or Material Palladium on 1/8-inch alumina pellets
Assay 0.5% (Pd)
Molecular Formula Pd
Formula Weight 106.42
Accessories

SDS & Safety Documents