missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 1% on 1-2 mm alumina spheres, reduced

Manufacturer:  Alfa Aesar™ 4482036

 View more versions of this product

Catalog No. AA4482036



Specifications

Specifications

Palladium
7440-05-3
MFCD03613602
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Sperical Powder
1 to 2mm, Reduced
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
500g

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03613602
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents