Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 0.5% on granular carbon, reduced

Hydrogenation of olefins

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 0.5% on granular carbon, Reduced
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3828914

  SDS
alfa aesar™
3828914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3828922

  SDS
alfa aesar™
3828922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 0.5% on granular carbon, Reduced
MDL Number MFCD03457879
Quantity 25g
Mesh Size 4x8 Mesh
Surface Area 900 to 1100m2/g