Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 0.5% on 3.18mm (0.125in) alumina pellets, reduced

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 0.5% on 3.18mm (0.125 in.) alumina pellets, Reduced
MDL Number MFCD03613602
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3878614

  SDS
alfa aesar™
3878614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3878622

  SDS
alfa aesar™
3878622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3878636

  SDS
alfa aesar™
3878636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 0.5% on 3.18mm (0.125 in.) alumina pellets, Reduced
MDL Number MFCD03613602
Quantity 25g
Odor Odorless