Please login to your online account to display your discounted pricing

Palatinose, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  0210255205

 View more versions of this product

Catalog No. ICN10255205

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical State Solid
Quantity 5g
Chemical Name or Material Palatinose
CAS 13718-94-0
Physical Form White Powder
Molecular Formula C12H22O1
Formula Weight 342.3g/mol
Synonym 6-O-α-D-Glucopyranosyl-D-fructose
Recommended Storage Store at room temperature
EINECS Number 237-282-1

Specifications

Chemical Name or Material Palatinose
CAS 13718-94-0
Physical Form White Powder
Molecular Formula C12H22O1
Formula Weight 342.3g/mol
Synonym 6-O-α-D-Glucopyranosyl-D-fructose
Accessories

Store at room temperature