Please login to your online account to display your discounted pricing

Palatinose, MP Biomedicals™

$166.55 - $166.55

Chemical Identifiers

Chemical Name or Material Palatinose
CAS 13718-94-0
Physical Form White Powder
Molecular Formula C12H22O1
Formula Weight 342.3g/mol
Synonym 6-O-α-D-Glucopyranosyl-D-fructose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10255205

 
mp biomedicals
0210255205
5g Each for $166.55
Description & Specifications

Specifications

Physical State Solid
Quantity 5g
Chemical Name or Material Palatinose
CAS 13718-94-0
Physical Form White Powder
Molecular Formula C12H22O1
Formula Weight 342.3g/mol
Synonym 6-O-α-D-Glucopyranosyl-D-fructose
Recommended Storage Store at room temperature
EINECS Number 237-282-1
Safety and Handling

Store at room temperature