SGE™ PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle
Features Diamond Syringe Technology which results in improved solvent resistance, wide temperature ranges, and increased operational smoothness. SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle are used for CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus, AS2000, AS3000, AS200/800 Autosamplers.
$79.46 - $581.72
Specifications
Type Autosampler Syringe
Connection Type Fixed
Needle Point Style Cone
For Use With (Application) Gas chromatography
Length (Metric) Needle 50 mm
View More Specs
Products 13
Catalog Number Mfr. No. Description Volume (Metric) Diameter (Metric) Outer Needle Needle Gauge Price Quantity  
Catalog Number Mfr. No. Description Volume (Metric) Diameter (Metric) Outer Needle Needle Gauge Price Quantity  
SG001981
View Documents
SGE™
SG001981
Not gas tight 5 µL 0.63 mm 23 G
Each for $145.61
SG002700
View Documents
SGE™
SG002700
Not gas tight 10 µL 0.47 mm 26 G
Each for $182.22
03-351-102
View Documents
SGE™
SG002967
Not gas tight 10 µL 0.5 mm
Each for $106.14
03-350-172
View Documents
SGE™
SG002976
Gas tight 10 µL 0.47 mm 26 G
Pack of 6 Each for $581.72
03-350-173
View Documents
SGE™
SG'002977
Gas tight 10 µL 0.47 mm 26 G
Each for $97.50
SG-002980
View Documents
SGE™
SG002980
Not gas tight 10 µL 0.47 mm 26 G
Each for $79.46
SG-002981
View Documents
SGE™
SG002981
Not gas tight 10 µL 0.63 mm 23 G
Each for $102.81
SG002986
View Documents
SGE™
SG002986
Not gas tight 10 µL 0.47 mm 26 G
Pack of 6 Each for $414.83
SG002987
View Documents
SGE™
SG002987
Gas tight 10 µL 0.63 mm 23 G
Each for $93.20
SG003700
View Documents
SGE™
SG003700
Gas tight 25 µL 0.47 mm 26 G
Each for $236.19
SG003987
View Documents
SGE™
SG003987
Gas tight 25 µL 0.63 mm 23 G
Each for $158.70
03-351-120
View Documents
SGE™
SG0039871
Gas tight 25 µL 0.63 mm 23 G
Each for $141.67
SG005335
View Documents
SGE™
SG005335
Gas tight 100 µL 0.63 mm 23 G
Each for $148.46
Description
Description
  • Improved solvent resistance
  • Wide temperature ranges up to 150°C
  • Increased operational smoothness
  • Designed specifically for the PAL
  • 22 gauge needles are 0.72mm OD
  • 23 gauge needles are 0.63mm OD
  • 26 gauge needles are 0.47mm OD
  • Accuracy and Reproducibility;  ±1% (dispensed volume) (±2% for 0.5μL syringes)
  • Cone shaped (also known as point style AS for autosampler), are designed to withstand multi injection demands and improves septum lifetime when used with an autosampler
  • Gas tight plunger, where applicable, is made of PTFE
Specifications
Specifications
Autosampler Syringe
Cone
50 mm
Fixed
Gas chromatography