Invitrogen™ PAK3 Recombinant Human Protein DFS Item
PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling.
Manufacturer:  Invitrogen™ LSPV4031
 View more versions of this product
Catalog No. PV4031
Description
Description
PAK proteins, a family of serine/threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities.
Specifications
Specifications
NP_002569
Frozen
68.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE20 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PAK3, bPAK, hPAK3
Baculovirus
Human
His Tag
Documents