missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ PAK2 (PAK65) Recombinant Human Protein
PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling.
Supplier: Invitrogen™ PV4566
Description
PAK proteins, a family of serine/threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities.
Specifications
NP_002568.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
PAK2 (PAK65), PAK6, PAK delta | |
Human | |
GST Tag | |
Frozen |
Nuclear Receptor Assay | |
85.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |