Please login to your online account to display your discounted pricing

PAH Mixture (For EPA 8100/Appendix IX)

$113.71 - $113.71

Chemical Identifiers

Chemical Name or Material PAH Mixture
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-PM-8101

 
ultra scientific
PM8101
Ampule 1x1mL Each for $113.71
Description & Specifications

Specifications

Quantity 1x1mL
Methods For EPA Method 8100/Appendix IX
Concentration or Composition (by Analyte or Components) Each component in Methylene Chloride:
Acenaphthene    •1000μg/mL
Acenaphthylene    •1000μg/mL
Anthracene    •1000μg/mL
Benz[a]anthracene    •100μg/mL
Benzo[b]fluoranthene    •100μg/mL
Benzo[k]fluoranthene    •50μg/mL
Benzo[ghi]perylene    •100μg/mL
Benzo[a]pyrene    •100μg/mL
Chrysene    •100μg/mL
Dibenz[a,h]anthracene    •100μg/mL
Fluoranthene    •100μg/mL
Fluorene    •1000μg/mL
Indeno[1,2,3-cd]pyrene    •100μg/mL
Naphthalene    •1000μg/mL
Phenanthrene    •1000μg/mL
Pyrene    •100μg/mL
Packaging Ampule
Chemical Name or Material PAH Mixture