Please login to your online account to display your discounted pricing
Thermo Scientific™ PageRuler™ Unstained High Range Protein Ladder
A mixture of 8 unstained proteins (60 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
$19.00 - $195.84
Specifications
Description PageRuler Unstained High Range Protein Ladder
Detection Visible
Formulation 8 proteins from 60 to 250kDa in reducing sample buffer
Stained Unstained
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    
PI26637
thermo scientific™
26637
Each for $195.84
PI26638
thermo scientific
26638
Each for $19.00
Description & Specifications
Specifications
Description PageRuler Unstained High Range Protein Ladder
Detection Visible
Formulation 8 proteins from 60 to 250kDa in reducing sample buffer
Stained Unstained
The Thermo Scientific™ PageRuler Unstained High Range Protein Ladder is a mixture of 8 unstained proteins (60 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
The protein MW markers in this ladder resolve into clearly identifiable, sharp bands when analyzed by SDS-PAGE and stained with coomassie dye or silver. The 150kDa band is of greater intensity and serves as a reference band. Proteins can be detected on Western blots by staining with Ponceau S, coomassie blue or other protein stains. In addition, the ladder proteins contain an integral Step-tag™ II Sequence and may be detected on Western blots using Strep-Tactin™ Conjugates. The PageRuler Unstained High Range Protein Ladder is supplied ready-to-use: no heating, dilution or addition of a reducing agent is required before use.
Highlights:
  • Size range 8 proteins spanning 60 to 250kDa
  • Ready-to-use supplied in a loading buffer for direct loading on gels
  • Sharp bands excellent accuracy for detection with coomassie or silver stains
  • Reference band 150kDa band has greater intensity for easy orientation
  • Tagged each protein in the ladder contains an integral Strep-tag™ II Sequence
  • Quality tested each lot evaluated by SDS-PAGE and Western blotting
Includes:
Proteins (0.02 to 0.05mg/mL of each) in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 100mM DTT, 1mM NaN3, 0.01% bromophenol blue and 33% glycerol
Recommended for:
Accurate protein sizing on SDS-polyacrylamide gels and Western blots