Thermo Scientific PageRuler™ Unstained High Range Protein Ladder
A mixture of 8 unstained proteins (60 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Manufacturer:  Thermo Scientific 26638
 View more versions of this product
Catalog No. PI26638
Specifications
Description
The Thermo Scientific™ PageRuler Unstained High Range Protein Ladder is a mixture of 8 unstained proteins (60 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
The protein MW markers in this ladder resolve into clearly identifiable, sharp bands when analyzed by SDS-PAGE and stained with coomassie dye or silver. The 150kDa band is of greater intensity and serves as a reference band. Proteins can be detected on Western blots by staining with Ponceau S, coomassie blue or other protein stains. In addition, the ladder proteins contain an integral Step-tag™ II Sequence and may be detected on Western blots using Strep-Tactin™ Conjugates. The PageRuler Unstained High Range Protein Ladder is supplied ready-to-use: no heating, dilution or addition of a reducing agent is required before use.
Highlights:
  • Size range 8 proteins spanning 60 to 250kDa
  • Ready-to-use supplied in a loading buffer for direct loading on gels
  • Sharp bands excellent accuracy for detection with coomassie or silver stains
  • Reference band 150kDa band has greater intensity for easy orientation
  • Tagged each protein in the ladder contains an integral Strep-tag™ II Sequence
  • Quality tested each lot evaluated by SDS-PAGE and Western blotting
Includes:
Proteins (0.02 to 0.05mg/mL of each) in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 100mM DTT, 1mM NaN3, 0.01% bromophenol blue and 33% glycerol
Recommended for:
Accurate protein sizing on SDS-polyacrylamide gels and Western blots
Description & Specifications
Specifications
25μL
Unstained
High
Visible
PageRuler Unstained High Range Protein Ladder
8 proteins from 60 to 250kDa in reducing sample buffer
2 to 5 mini gel lanes at 5 to 10uL per lane