Please login to your online account to display your discounted pricing

Alfa Aesar™ PACAP-Related Peptide, Human

For cellular and molecular biology applications

Specifications

Product Type PACAP-Related Peptide, human
Cross Reactivity Human
Molecular Weight 3146.60
Storage Requirements -30° to -10°C
Sequence Asp-Val-Ala-His-Gly-Ile-Leu-Asn-Glu-Ala-Tyr-Arg-Lys-Val-Leu-Asp-Gln-Leu-Ser-Ala-Gly-Lys-His-Leu-Gln-Ser-Leu-Val-Ala
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66104EXD

 
alfa aesar™
J66104EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66104MCR

 
alfa aesar™
J66104MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type PACAP-Related Peptide, human
Cross Reactivity Human
Molecular Weight 3146.60
Storage Requirements -30° to -10°C
Sequence Asp-Val-Ala-His-Gly-Ile-Leu-Asn-Glu-Ala-Tyr-Arg-Lys-Val-Leu-Asp-Gln-Leu-Ser-Ala-Gly-Lys-His-Leu-Gln-Ser-Leu-Val-Ala
Format Solid
Common Name Pituitary adenylate cyclase activating polypeptide-related peptide
Regulatory Status RUO
Purification Method HPLC