Please login to your online account to display your discounted pricing

Alfa Aesar™ PACAP (6-27), Human, Ovine, Rat

For cellular and molecular biology applications

Specifications

Product Type PACAP (6-27), human, ovine, rat
Cross Reactivity Human, Ovine, Rat
Molecular Weight 2638.13
Storage Requirements -30° to -10°C
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66872EXD

 
alfa aesar™
J66872EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66872MCR

 
alfa aesar™
J66872MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type PACAP (6-27), human, ovine, rat
Cross Reactivity Human, Ovine, Rat
Molecular Weight 2638.13
Storage Requirements -30° to -10°C
Format Solid
Sequence Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2
Regulatory Status RUO
Purification Method HPLC