Please login to your online account to display your discounted pricing

Alfa Aesar™ PACAP (1-38), Human, Ovine, Rat

For cellular and molecular biology applications

Specifications

Product Type PACAP (1-38), human, ovine, rat
Molecular Weight 4534.31
Storage Requirements -30° to -10°C
Format Lyophilized solid
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66444EXD

 
alfa aesar™
J66444EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66444MCR

 
alfa aesar™
J66444MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type PACAP (1-38), human, ovine, rat
Molecular Weight 4534.31
Storage Requirements -30° to -10°C
Format Lyophilized solid
Regulatory Status RUO
Purification Method HPLC