Honeywell™ North™ Primair™ PA700 PAPR Kit
Offers convenience, comfort, confidence, and connectivity.
$1,880.13 - $2,234.34
Specifications
Product Type PAPR Kit
View More Specs
Products 2
Catalog Number Mfr. No. Includes Assigned Protection Factor (APF) Price Quantity  
Catalog Number Mfr. No. Includes Assigned Protection Factor (APF) Price Quantity  
19-085-544
View Documents Promotion Details
Honeywell™
PA701HED101TF
Medium Sized loose-fitting head cover and adjustable headgear 25
Each for $2,234.34
19-085-543
View Documents Promotion Details
Honeywell™
PA701HED111TF
Universal size bibbed hood 1000
Each for $1,880.13
Description
Description
The PA700 Series PAPR is easy to assembly with components that lock into place with little effort. Controls are easy to operate—even when wearing gloves. Easy-to-clean, smooth surfaces take just minutes to wipe down at the end of a shift.
  • Blower includes intelligent three-alarm system (visual, audible, and vibratory) to clearly alert users of battery and airflow status
  • Illuminated keypad features easy-to-read display and large buttons for convenient operation
  • On-demand airflow with three speeds enhance user comfort
  • Smart breathing tube connection automatically detects between tight and loose fitting hoses and adjusts airflow accordingly
  • Lightweight flexible breathing tube provides increased freedom of movement
  • Ergonomic belt distributes PAPR weight evenly and prevents slippage
  • Adjustable easy-to-clean PVC straps provide customized fit
  • Side-mounted battery locks into place to reduce accidental disconnects
  • Powered by rechargeable lithium-ion battery pack with on-board charge display; battery holds 8-hour charge
  • Smart-charger system can re-charge fully depleted battery in approximately 4.5 hours; continuously monitors battery pack health and alerts user of potential problems via the LED display panel
  • Assigned Protection Factor: 25 APF for loose-fitting head covering; 1000 APF for bibbed hood
  • Weight, 6.0 lb. (2.7kg)
Compliance
Meets the requirements of NIOSH CFR 84 when assembled from components listed on the NIOSH approval label included with the product.
Specifications
Specifications
PAPR Kit