Please login to your online account to display your discounted pricing

P7C3, Tocris™

Neuroprotective compound

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material P7C3
Assay Percent Range >99%
Molecular Formula C21H18Br2N2O
Formula Weight 474.19
Synonym 3, 6-Dibromo-α-[(phenylamino)methyl]-9H-carbazole-9-ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

407610

 
r & d systems
407610
10mg Each for $244.81

407650

 
r & d systems
407650
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 474.19
Quantity 10mg
Molecular Formula C21H18Br2N2O
CAS 301353-96-8
Chemical Name or Material P7C3
Assay Percent Range >99%
Synonym 3, 6-Dibromo-α-[(phenylamino)methyl]-9H-carbazole-9-ethanol
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature