Please login to your online account to display your discounted pricing

Alfa Aesar™ P75-TNFR

For cellular and molecular biology applications

Specifications

Product Type P75-TNFR
Molecular Weight 1204.40
Storage Requirements -30° to -10°C
Sequence Ser-Met-Ala-Pro-Gly-Ala-Val-His-Leu-Pro-Gln-Pro
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66617LB0

 
alfa aesar™
J66617LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66617MA

 
alfa aesar™
J66617MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type P75-TNFR
Molecular Weight 1204.40
Storage Requirements -30° to -10°C
Sequence Ser-Met-Ala-Pro-Gly-Ala-Val-His-Leu-Pro-Gln-Pro
Format Solid
Common Name TNFR-II
Regulatory Status RUO
Purification Method HPLC