Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p57 Kip2 Ab-7, Rabbit Polyclonal Antibody


Deliver accurate, reproducible results with the Thermo Scientific™ p57Kip2 Ab-7, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1637P1ABX

 View more versions of this product

Catalog No. RB1637P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 500µL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Host Species Rabbit
Immunogen A synthetic peptide from human p57superscriptKip2superscript protein
Regulatory Status RUO
Antigen p57superscriptKip2superscript Ab-7
Applications Immunohistochemistry (Paraffin)
Format Concentrated
Type Primary
Classification Polyclonal
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cancer and Tumor Biology
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 500uL; 1mg/mL; Purified w/o BSA, azide

Deliver accurate, reproducible results with the Thermo Scientific™ p57Kip2 Ab-7, Rabbit Polyclonal Antibody.

p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Cow and Mouse. Others not known.

Epitope: Near C-terminus

Immunogen: A synthetic peptide from human p57Kip2 protein

Molecular Weight: 57kDa

Positive Control: Colon CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)

Note:

USA: RUO; Int'l: RUO