Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p53 Ab-6, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-6, Mouse Monoclonal Antibody.

$243.39 - $1,045.88

Specifications

Antigen p53 Ab-6
Clone DO-1
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS187P

 
thermo scientific™ lab vision™
MS187P
Purified Ab with BSA and Azide 1mL Each for $969.90

MS187P0

 
thermo scientific™ lab vision™
MS187P0
Purified Ab with BSA and Azide 100µL Each for $243.39

MS187P1

 
thermo scientific™ lab vision™
MS187P1
Purified Ab with BSA and Azide 500µL Each for $677.94

MS187P1ABX

 
thermo scientific™ lab vision™
MS187P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $758.24

MS187PABX

 
thermo scientific™ lab vision™
MS187PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,045.88
Description & Specifications

Specifications

Antigen p53 Ab-6
Clone DO-1
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Recombinant human wild-type p53 protein expressed in E. coli
Isotype IgG2a

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-6, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-1

Isotype: IgG2a

Species Reactivity: Human, Monkey and Cow. Reacts weakly with Mouse and Rat. Others not known.

Epitope: aa 11-25

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 cells or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO