missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p53 Ab-6, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-6, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS187P1ABX

 View more versions of this product

Catalog No. MS187P1ABX


 

Specifications

Description

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-6, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-1

Isotype: IgG2a

Species Reactivity: Human, Monkey and Cow. Reacts weakly with Mouse and Rat. Others not known.

Epitope: aa 11-25

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 cells or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

p53 Ab-6
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG2a
RUO
Primary
Bovine,Human,Non-human Primate
Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
DO-1
Unlabeled
Concentrated
Recombinant human wild-type p53 protein expressed in E. coli
100μL
Cell Cycle
Monoclonal