Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p53 Ab-2, Mouse Monoclonal Antibody


Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS105P1ABX

 View more versions of this product

Catalog No. MS105P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen p53 Ab-2
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone PAb 1801
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Recombinant Human p53 protein
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-2, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 1801

Isotype: IgG1

Species Reactivity: Human specific.

Epitope: aa 32-79

Immunogen: Recombinant human p53 protein

Molecular Weight: 53kDa

Positive Control: SKBR-3 cells, MDA-231 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO