Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p53 Ab-1, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-1, Mouse Monoclonal Antibody.

$244.84 - $1,163.51

Specifications

Antigen p53 Ab-1
Clone PAb 240
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS104P

 
thermo scientific™ lab vision™
MS104P
Purified Ab with BSA and Azide 1mL Each for $961.94

MS104P0

 
thermo scientific™ lab vision™
MS104P0
Purified Ab with BSA and Azide 100µL Each for $244.84

MS104P1

 
thermo scientific™ lab vision™
MS104P1
Purified Ab with BSA and Azide 500µL Each for $682.04

MS104P1ABX

 
thermo scientific™ lab vision™
MS104P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $753.96

MS104PABX

 
thermo scientific™ lab vision™
MS104PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,163.51

MS104R7

 
thermo scientific™ lab vision™
MS104R7
Ready-to-use for IHC (UVLP) 7mL Each for $606.63
Description & Specifications

Specifications

Antigen p53 Ab-1
Clone PAb 240
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Murine p53 protein corresponding to aa14-289
Isotype IgG1

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-1, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 240

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 212-217

Immunogen: Murine p53 protein corresponding to aa 14-289

Molecular Weight: 53kDa

Positive Control: MDA-231 or SKBR-3 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Methacarn or formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO