missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p300/CBP (CREB-Binding Protein) Ab-1 Mouse Monoclonal Antibody, Epredia™
Achieve accurate, reproducible results with the Epredia™ p300/CBP (CREB-Binding Protein) Ab-1, Mouse Monoclonal Antibody.
$1123.02 - $2181.69
Specifications
Antigen | p300/CBP (CREB-Binding Protein) Ab-1 |
---|---|
Clone | NM11 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity | |||
MS586B1
|
Thermo Fisher Scientific
MS586B1 |
Concentrated | Biotin | 500 μL |
Each for $1,463.94
|
|
|||
MS586B
|
Thermo Fisher Scientific
MS586B |
Concentrated | Biotin | 1 mL |
Each for $2,181.69
|
|
|||
MS586P1ABX
|
Thermo Fisher Scientific
MS586P1ABX |
Concentrated | Unconjugated | 100 μL |
Each for $1,297.71
|
|
|||
MS586PABX
|
Thermo Fisher Scientific
MS586PABX |
Concentrated | Unconjugated | 200 μL |
Each for $1,900.23
|
|
|||
MS586P1
|
Thermo Fisher Scientific
MS586P1 |
Concentrated | Unconjugated | 500 μL |
Each for $1,139.00
|
|
|||
MS586R7
|
Thermo Fisher Scientific
MS586R7 |
Prediluted | Unconjugated | 7 mL |
Each for $1,123.02
|
|
|||
Description
p300 is a transcriptional adapter protein which is characterized by three cysteine- and histidine-rich regions and its C-terminus specifically binds the adenovirus E1A protein. p300, and p300-associated proteins, are components of TATA-binding protein (TBP) complexes. Protein kinase A mediated CREB phosphorylation results in the binding of CREB to a 265kDa nuclear protein designated CBP (for CREB-binding protein). CBP and p300 are homologous to each other.Host Species: Mouse
Clone: NM11
Isotype: IgG2b/κ
Species Reactivity: Human, Monkey, Mink, Mouse, and Rat. Others not known.
Epitope: aa 2071-2091
Immunogen: E1A affinity purified p300 protein
Molecular Weight: 300kDa
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
p300/CBP (CREB-Binding Protein) Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Mouse | |
Transcription Regulation | |
E1A affinity purified p300 protein | |
Primary |
NM11 | |
Monoclonal | |
RUO | |
Human, Mink, Mouse, Non-human Primate, Rat | |
IgG2b κ |