Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p27 Kip1 Ab-1, Mouse Monoclonal Antibody


Achieve accurate, reproducible results with the Thermo Scientific™ p27Kip1 Ab-1, Mouse Monoclonal Antibody. 

Manufacturer: thermo scientific™ lab vision™  MS256P1ABX

 View more versions of this product

Catalog No. MS256P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen p27superscriptKip1superscript Ab-1
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone DCS-72.F6
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Immunogen Mouse recombinant p27 protein
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Canine
Target Species Human
Target Species Murine
Target Species Rat

Achieve accurate, reproducible results with the Thermo Scientific™ p27Kip1 Ab-1, Mouse Monoclonal Antibody.

p27Kip1, a candidate tumor suppressor gene, functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF-Β induced G1 arrest.

Host Species: Mouse

Clone: DCS-72.F6

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, and Dog. Does not react with cow. Others not known.

Epitope: aa 83-197

Immunogen: Mouse recombinant p27 protein

Molecular Weight: 27kDa

Positive Control: MAD109, Ls174T cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO