Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p21 WAF1 Ab-5, Mouse Monoclonal Antibody


Achieve accurate, reproducible results with the Thermo Scientific™ p21WAF1 Ab-5, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS387P1ABX

 View more versions of this product

Catalog No. MS387P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Host Species Mouse
Immunogen Full length human recombinant p21 protein
Isotype IgG1
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Clone HZ52
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Rat
Quantity 100µL
Antigen p21superscriptWAF1superscript Ab-5
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Research Discipline Cancer and Tumor Biology
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Achieve accurate, reproducible results with the Thermo Scientific™ p21WAF1 Ab-5, Mouse Monoclonal Antibody.

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: HZ52

Isotype: IgG1

Species Reactivity: Human, Cow, Mouse, and Rat. Others not known.

Epitope: Not determined

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Ls174T or Raji cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO