Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p21 WAF1 Ab-11, Mouse Monoclonal Antibody

Ensure accurate, reproducible results with the Thermo Scientific™ p21WAF1 Ab-11, Mouse Monoclonal Antibody.

$290.82 - $1,530.92

Specifications

Antigen p21superscriptWAF1superscript Ab-11
Clone CP74
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS891B

 
thermo scientific™ lab vision™
MS891B
200μg/mL Biotin 1mL Each for $1,530.92

MS891B0

 
thermo scientific™ lab vision™
MS891B0
200μg/mL Biotin 100µL Each for $389.35

MS891B1

 
thermo scientific™ lab vision™
MS891B1
200μg/mL Biotin 500µL Each for $1,036.02

MS891P

 
thermo scientific™ lab vision™
MS891P
200μg/mL Unlabeled 1mL Each for $1,195.61

MS891P0

 
thermo scientific™ lab vision™
MS891P0
200μg/mL Unlabeled 100µL Each for $290.82

MS891P1

 
thermo scientific™ lab vision™
MS891P1
200μg/mL Unlabeled 500µL Each for $780.31

MS891P1ABX

 
thermo scientific™ lab vision™
MS891P1ABX
1mg/mL Unlabeled 100µL Each for $873.94

MS891PABX

 
thermo scientific™ lab vision™
MS891PABX
1mg/mL Unlabeled 200µL Each for $1,287.43
Description & Specifications

Specifications

Antigen p21superscriptWAF1superscript Ab-11
Clone CP74
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Full length human recombinant p21 protein
Isotype IgG2b

Ensure accurate, reproducible results with the Thermo Scientific™ p21WAF1 Ab-11, Mouse Monoclonal Antibody.

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: CP74

Isotype: IgG2b

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-80

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Raji, PC12 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO