Please login to your online account to display your discounted pricing

Thermo Scientific™ p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1, Mouse Monoclonal Antibody

Ensure provides accurate, reproducible results with the Thermo Scientific™ p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1, Mouse Monoclonal Antibody.

$275.39 - $1,200.67

Specifications

Antigen p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1
Clone 265/F4
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS190P

 
thermo scientific™
MS190P
200μg/mL 1mL Each for $1,126.88

MS190P0

 
thermo scientific™
MS190P0
200μg/mL 100µL Each for $275.39

MS190P1

 
thermo scientific™
MS190P1
200μg/mL 500µL Each for $739.80

MS190P1ABX

 
thermo scientific™
MS190P1ABX
1mg/mL 100µL Each for $837.33

MS190PABX

 
thermo scientific™
MS190PABX
1mg/mL 200µL Each for $1,200.67
Description & Specifications

Specifications

Antigen p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1
Clone 265/F4
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Hamster
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Colchicine-resistant viable Chinese Hamster Ovary (CHO) cells, followed by purified P170 glycoprotein from CHO cells
Isotype IgG2a kappa

Ensure provides accurate, reproducible results with the Thermo Scientific™ p170/p-Glycoprotein/MDR (Anti-Hamster) Ab-1, Mouse Monoclonal Antibody.

Multi-drug resistance(MDR) associated p-glycoprotein (p170) causes resistance to various drugs in a number of cancer patients. It exists in a soluble and membrane associated form. Soluble P-glycoprotein is detected in extracellular fluids of cancer patients, such as malignant ascites and serum. The Mr of soluble P-glycoprotein is the same as that of membrane bound P-glycoprotein.

Host Species: Mouse

Clone: 265/F4

Isotype: IgG1/κ

Species Reactivity: Chinese Hamster. Does not react with human. Others not known.

Epitope: Not determined

Immunogen: Colchicine-resistant viable Chinese Hamster Ovary (CHO) cells, followed by purified p170 glycoprotein from CHO cells

Molecular Weight: 170kDa

Positive Control: Drug-resistant Chinese Hamster Ovary (CHO) cells

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO