missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ p170/p-Glycoprotein/MDR Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results with the Thermo Scientific™ p170/p-Glycoprotein/MDR Ab-2, Mouse Monoclonal Antibody.

$244.18 - $1,207.00

Specifications

Antigen p170/p-Glycoprotein/MDR Ab-2
Clone F4
Applications Immunofluorescence,Western Blotting,Immunofluorescence,Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS660P Thermo Scientific™ Lab Vision™
MS660P
200μg/mL 1mL Each for $1,120.63
MS660P0 Thermo Scientific™ Lab Vision™
MS660P0
200μg/mL 100μL Each for $244.18
MS660P1 Thermo Scientific™ Lab Vision™
MS660P1
200μg/mL 500μL Each for $713.80
MS660P1ABX Thermo Scientific™ Lab Vision™
MS660P1ABX
1mg/mL 100μL Each for $795.22
MS660PABX Thermo Scientific™ Lab Vision™
MS660PABX
1mg/mL 200μL Each for $1,207.00
Specifications

Description

Ensure accurate, reproducible results with the Thermo Scientific™ p170/p-Glycoprotein/MDR Ab-2, Mouse Monoclonal Antibody.

Multi-drug resistance(MDR) associated p-glycoprotein (p170) causes resistance to various drugs in a number of cancer patients. It exists in a soluble and membrane associated form. Soluble P-glycoprotein is detected in extracellular fluids of cancer patients, such as malignant ascites and serum. The Mr of soluble P-glycoprotein is the same as that of membrane bound P-glycoprotein.

Host Species: Mouse

Clone: F4

Isotype: IgG1

Species Reactivity: Human, Chinese Hamster. Does not react with mouse. Others not known.

Epitope: Extracellular amino terminal half. This epitope is resistant to formalin fixation and periodate oxidation and is different from those of other commonly known MAbs to p-glycoprotein, such as C219, JSB-1, and MRK16, which recognize an intracellular and extracellular epitope of p-glycoprotein, respectively.

Immunogen: Human and hamster drug-resistant viable cells followed by crude plasma membranes

Molecular Weight: 170kDa

Positive Control: Drug-resistant cells, Kidney or breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry (Fixed cells only)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

p170/p-Glycoprotein/MDR Ab-2
Immunofluorescence,Western Blotting,Immunofluorescence,Western Blotting
Unlabeled
Murine
Cancer and Tumor Biology
Human and hamster drug-resistant viable cells followed by crude plasma membranes
F4
Monoclonal
Concentrated
RUO
Primary
IgG1