Please login to your online account to display your discounted pricing

P1075, Tocris

Potent Kir6 (KATP) channel opener

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material P1075
Assay Percent Range >99%
Molecular Formula C12H17N5
Formula Weight 231.3
Synonym N-cyano-N'-(1, 1-dimethylpropyl)-N''-3-pyridylguanidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

135550

 
r & d systems
RDS135550
50mg Each for $976.15

135510

 
r & d systems
RDS135510
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 231.3
Quantity 50mg
Chemical Name or Material P1075
Assay Percent Range >99%
Molecular Formula C12H17N5
CAS 60559-98-0
Synonym N-cyano-N'-(1, 1-dimethylpropyl)-N''-3-pyridylguanidine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature