Honeywell™ North™ P100 HEPA Filter
Designed for use with Honeywell North Primair PA700 Series Powered Air Purifying Respirator (PAPR).
Supplier: Honeywell™ PA7HE
Description
- Filter locks into place with a quarter turn, assuring workers they are secure
- Provides 99.97% minimum filter efficiency for all particulates
Specifications
HEPA | |
Filter | |
PA700 |