missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Xylylenebis(triphenylphosphonium chloride), 97+%

$81.58 - $268.32

Chemical Identifiers

CAS 1519-47-7
Molecular Formula C44H38Cl2P2
Molecular Weight (g/mol) 699.636
MDL Number MFCD00031710
InChI Key IUNJPQCPINLADS-UHFFFAOYSA-L
Synonym p-xylylenebis triphenylphosphonium chloride, 1,4-phenylenebis methylene bis triphenylphosphonium chloride, phosphonium, p-xylylenebis triphenyl-, dichloride, p-phenylenebis methylene bis triphenylphosphonium dichloride, phosphonium, p-phenylenedimethylene bis triphenyl-, dichloride, phosphonium, 1,4-phenylenebis methylene bis triphenyl-, dichloride, phosphonium, 1,4-phenylenebis methylene bis triphenyl-, dichloride, 1,4-phenylenebis methylene bis triphenylphosphonium dichloride, xylylenebistriphenylphosphoniumchloride, p-xylylenebis triphenylphosphonium chloride 5g
PubChem CID 168940
IUPAC Name triphenyl-[[4-(triphenylphosphaniumylmethyl)phenyl]methyl]phosphanium;dichloride
SMILES C1=CC=C(C=C1)[P+](CC2=CC=C(C=C2)C[P+](C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7.[Cl-].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0133406 View Documents Alfa Aesar™
L0133406
5g Each for $81.58
 
AAL0133414 View Documents Alfa Aesar™
L0133414
25g Each for $268.32
 
Specifications

Chemical Identifiers

1519-47-7
699.636
IUNJPQCPINLADS-UHFFFAOYSA-L
168940
C1=CC=C(C=C1)[P+](CC2=CC=C(C=C2)C[P+](C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7.[Cl-].[Cl-]
C44H38Cl2P2
MFCD00031710
p-xylylenebis triphenylphosphonium chloride, 1,4-phenylenebis methylene bis triphenylphosphonium chloride, phosphonium, p-xylylenebis triphenyl-, dichloride, p-phenylenebis methylene bis triphenylphosphonium dichloride, phosphonium, p-phenylenedimethylene bis triphenyl-, dichloride, phosphonium, 1,4-phenylenebis methylene bis triphenyl-, dichloride, phosphonium, 1,4-phenylenebis methylene bis triphenyl-, dichloride, 1,4-phenylenebis methylene bis triphenylphosphonium dichloride, xylylenebistriphenylphosphoniumchloride, p-xylylenebis triphenylphosphonium chloride 5g
triphenyl-[[4-(triphenylphosphaniumylmethyl)phenyl]methyl]phosphanium;dichloride

Specifications

>300°C
1519-47-7
MFCD00031710
3876684
IUNJPQCPINLADS-UHFFFAOYSA-L
triphenyl-[[4-(triphenylphosphaniumylmethyl)phenyl]methyl]phosphanium;dichloride
168940
≥97%
5g
C44H38Cl2P2
UN3464
p-xylylenebis triphenylphosphonium chloride, 1,4-phenylenebis methylene bis triphenylphosphonium chloride, phosphonium, p-xylylenebis triphenyl-, dichloride, p-phenylenebis methylene bis triphenylphosphonium dichloride, phosphonium, p-phenylenedimethylene bis triphenyl-, dichloride, phosphonium, 1,4-phenylenebis methylene bis triphenyl-, dichloride, phosphonium, 1,4-phenylenebis methylene bis triphenyl-, dichloride, 1,4-phenylenebis methylene bis triphenylphosphonium dichloride, xylylenebistriphenylphosphoniumchloride, p-xylylenebis triphenylphosphonium chloride 5g
C1=CC=C(C=C1)[P+](CC2=CC=C(C=C2)C[P+](C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7.[Cl-].[Cl-]
699.636
699.66
p-Xylylenebis(triphenylphosphonium chloride)
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.

RTECSNumber : TA3580000

TSCA : TSCA