missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Xylylenebis(triphenylphosphonium bromide), 96%

$65.68 - $179.51

Chemical Identifiers

CAS 40817-03-6
Molecular Formula C44H38Br2P2
Formula Weight 788.55
MDL Number MFCD00011914
Beilstein 3584592
Chemical Name or Material p-Xylylenebis(triphenylphosphonium bromide)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1824114 SDS Alfa Aesar™
A1824114
25g Each for $65.68
AAA1824122 SDS Alfa Aesar™
A1824122
100g Each for $179.51
Specifications

Description

Applications
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

40817-03-6
788.55
3584592
C44H38Br2P2
MFCD00011914
p-Xylylenebis(triphenylphosphonium bromide)

Specifications

>300°C
40817-03-6
788.55
3584592
96%
25g
C44H38Br2P2
MFCD00011914
p-Xylylenebis(triphenylphosphonium bromide)

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning