Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Xylene-d{10}, 99%(Isotopic)

Chemical Identifiers

Chemical Name or Material p-Xylene-d{10}
CAS 41051-88-1
Name Note Isotopic
Formula Weight 116.25
MDL Number MFCD00037699
UN Number UN1307
Merck Index 14,10081
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3647306

  SDS
alfa aesar™
3647306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.25
Chemical Name or Material p-Xylene-d{10}
Sensitivity Moisture Sensitive
Density 0.948g/mL
Name Note Isotopic
Boiling Point 137° to 138°C
Melting Point 12° to 13°C
Physical Form Liquid
Assay 99%
Quantity 5g
MDL Number MFCD00037699
CAS 41051-88-1
UN Number UN1307
Merck Index 14,10081
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H226-H332-H315
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H226-H332-H315
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.