Please login to your online account to display your discounted pricing

p-Xylene (Certified), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  O50824

 View more versions of this product

Catalog No. O5082-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Purity Grade Certified Grade
Boiling Point 138°C
Melting Point 13°C
Formula Weight 106.17
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Vapor Pressure 8mbar at 20°C
Vapor Density 3.7g/mol
Viscosity 0.648 mPa/s at 20°C
Alpha Vector Xylene
Color Colorless
Molecular Formula C8H10
Density 0.866g/cm³
Packaging Glass Bottle
Cleanup Symbol Solvent
MDL Number MFCD00008556
Identification Pass Test
Linear Formula C6H4(CH3)2
ChemAlert Storage Symbol Red
Chemical Name or Material p-Xylene
CAS 106-42-3

Specifications

Chemical Name or Material p-Xylene
CAS 106-42-3
Purity Grade Certified Grade
Assay ≥99%
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
Formula Weight 106.17
MDL Number MFCD00008556
Accessories

Accessories for Catalog Number: O50824

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
   

SDS & Safety Documents