missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Xylene, 99%, pure, ACROS Organics™

Manufacturer:  Acros Organics AC158770100

 View more versions of this product

Catalog No. AC158770100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

p-Xylene
99%
C6H4(CH3)2
05, 382
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
URLKBWYHVLBVBO-UHFFFAOYSA-N
1,4-xylene
7809
106.17
99%
0.8660g/mL
Plastic drum
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
25°C
138.0°C
13.0°C
106-42-3
C8H10
MFCD00008556
15, 10278
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
CC1=CC=C(C=C1)C
106.168
CHEBI:27417
Liquid
Pure
Authentic
1.4940 to 1.4960
0.866
0.648 mPa.s (20°C)
Colorless to Yellow
10L

Chemical Identifiers

106-42-3
106.168
URLKBWYHVLBVBO-UHFFFAOYSA-N
7809
1,4-xylene
C8H10
MFCD00008556
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
CHEBI:27417
CC1=CC=C(C=C1)C
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents