Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Tolylurea, 98+%

$1,310.88 - $1,310.88

Chemical Identifiers

Chemical Name or Material p-Tolylurea
CAS 622-51-5
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00025433
Beilstein 2208088
Synonym 4-Methylphenylurea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1115036

  SDS
alfa aesar™
A1115036
500g Each for $1,310.88

AAA1115014

  SDS
alfa aesar™
A1115014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1115022

  SDS
alfa aesar™
A1115022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material p-Tolylurea
Melting Point 177° to 184°C
Quantity 500g
CAS 622-51-5
RTECS Number YU4200000
Assay ≥98%
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00025433
Beilstein 2208088
Synonym 4-Methylphenylurea