missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Tolylurea, 98+%

Manufacturer:  Alfa Aesar™ A1115036

 View more versions of this product

Catalog No. AAA1115036



Specifications

Specifications

p-Tolylurea
500g
≥98%
MFCD00025433
4-Methylphenylurea
CC1=CC=C(C=C1)NC(=O)N
150.181
150.18
177°C to 184°C
622-51-5
C8H10N2O
2208088
DMSHKWHLXNDUST-UHFFFAOYSA-N
(4-methylphenyl)urea
12148

Chemical Identifiers

622-51-5
150.181
DMSHKWHLXNDUST-UHFFFAOYSA-N
12148
CC1=CC=C(C=C1)NC(=O)N
C8H10N2O
MFCD00025433
4-Methylphenylurea
(4-methylphenyl)urea
Safety and Handling

Safety and Handling

RTECSNumber : YU4200000

SDS Documents