Please login to your online account to display your discounted pricing

p-Tolylmagnesium chloride, 2M (31 wt.%) solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC382721000

 View more versions of this product

Catalog No. AC382721000

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Dark
Physical Form Solution
Formula Weight 150.89
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Concentration 2 to 2.2M
Density 0.9700g/mL
Chemical Name or Material p-Tolylmagnesium chloride
Name Note 2M solution in THF
Molecular Formula C7H7ClMg
Linear Formula CH3C6H4MgCl
MDL Number MFCD00672097
CAS 696-61-7
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.
CAS 109-99-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material p-Tolylmagnesium chloride
Name Note 2M solution in THF
CAS 696-61-7
CAS 109-99-9
Molecular Formula C7H7ClMg
Linear Formula CH3C6H4MgCl
Formula Weight 150.89
MDL Number MFCD00672097
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.

SDS & Safety Documents