Please login to your online account to display your discounted pricing

p-Tolylmagnesium chloride, 2M (31 wt.%) solution in THF, AcroSeal™, ACROS Organics™

$250.06 - $250.06

Chemical Identifiers

Chemical Name or Material p-Tolylmagnesium chloride
Name Note 2M solution in THF
CAS 696-61-7
CAS 109-99-9
Molecular Formula C7H7ClMg
Linear Formula CH3C6H4MgCl
Formula Weight 150.89
MDL Number MFCD00672097
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382721000

  SDS
thermo fisher scientific
AC382721000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $250.06

AC382728000

  SDS
thermo fisher scientific
AC382728000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 150.89
Color Dark
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Concentration 2 to 2.2M
Density 0.9700g/mL
Linear Formula CH3C6H4MgCl
Name Note 2M solution in THF
CAS 696-61-7
Molecular Formula C7H7ClMg
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.
CAS 109-99-9
Health Hazard 1 Danger
Chemical Name or Material p-Tolylmagnesium chloride
MDL Number MFCD00672097
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.