missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Tolyldiethanolamine 98.0+%, TCI America™
Supplier: TCI America T0797100G
Specifications
| p-Tolyldiethanolamine | |
| 52°C | |
| 163°C | |
| MFCD00020580 | |
| N,N-Bis(2-hydroxyethyl)-p-toluidine, 2,2′-(4-Methylphenylimino)diethanol, 2,2′-(p-Tolylimino)diethanol | |
| CC1=CC=C(C=C1)N(CCO)CCO | |
| 195.26 | |
| 195.26 | |
| Crystalline Powder |
| 3077-12-1 | |
| White-Yellow | |
| C11H17NO2 | |
| 100 g | |
| JUVSRZCUMWZBFK-UHFFFAOYSA-N | |
| 2-[N-(2-hydroxyethyl)-4-methylanilino]ethanol | |
| 76497 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3077-12-1 | |
| 195.26 | |
| JUVSRZCUMWZBFK-UHFFFAOYSA-N | |
| 2-[N-(2-hydroxyethyl)-4-methylanilino]ethanol |
| C11H17NO2 | |
| MFCD00020580 | |
| 76497 | |
| CC1=CC=C(C=C1)N(CCO)CCO |
Safety and Handling
EINECSNumber : (3)-0233
TSCA : Yes